Anti-IDH Rabbit Polyclonal Antibody
Catalog # AGRIAS06203A
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Isocitrate dehydrogenase
- Antigen symbol:IDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:R. communis
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide 1 and peptide 2 conserved in all higher plants mitochondrial, NAD dependent isocitrate dehydrogenase subunits including Arabidopsis thaliana IDH-I Q8LFC0, At4g35260 and IDH-II P93032, At2g17130
- Size:180 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: R. communis