Anti-PC Rabbit Polyclonal Antibody
Catalog # AGRIAS06141
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:purified native plastocyanin from Spinacia oleracea P00289
- Tested applications:Immuno-Gold
- Purification:purified
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human