Anti-PsbF Rabbit Polyclonal Antibody
AGRIAS06113
: Agrisera
- Antibody type:Primary
- Antigen symbol:PSBF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:T. pseudonana,Hordeum vulgare,Lotus corniculatus,Anabaena pcc 7120,Phaseolus vulgaris,Oryza sativa,Ulva prolifera,Trifolium pratense,C. reinhardtii,Synechocystis sp. pcc 6803,Synechococcus sp. pcc 7942,Pisum sativum,A. thaliana,N. tabacum,T. guillardii,Ditylum brightwellii
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from plant PsbF sequences including Arabidopsis thaliana P62095, AtCg00570
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: PSBF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Horderum vulgare, C. reinhardtii, Lotus corniculatus, N. tabacum, Oryza sativa, Pisum sativum, Phaseolus vulgaris, Trifolium pratense, Ulva prolifera, Synechococcus sp. PCC 7942, Synechocystis sp. PCC6803, Anabaena 7120, Ditylum brightwellii, T. guillardii, T. pseudonana