Anti-HSP70 Rabbit Polyclonal Antibody
AGRIAS05061
: Agrisera
- Antibody type:Primary
- Antigen symbol:HSP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide chosen from the C-terminal of salmonid hsp70. The target peptide is a sequence specific to salmonid hsp70 B5X4Z3.
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: A. thaliana