Anti-RbcL Chicken Polyclonal Antibody
Catalog # AGRIAS01017
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:RBCL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- Reactivity:Spinacia oleracea,Phytoplankton,Solanum tuberosum,Synechococcus sp. pcc 7842,Pisum sativum,A. thaliana,Lobaria pulmonaria,Medicago sativa,Thiobacillus sp. ulmus sp.,Spartina alterniflora
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:O03042
- Storage temperature:+4 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from all known plant,algal and cyanobacterial RbcL (Rubisco large subunit of Rubisco Form I) sequences, including Arabidopsis thaliana
- Tested applications:Immunolocalization
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RBCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: A. thaliana, Lobaria pulmonaria, Medicago sativa, Phytoplankton, Pisum sativum, Solanum tuberosum, Spartina alterniflora, Spinacia oleracea, Synechococcus sp. pcc7842, Thiobacillus sp. Ulmus sp.