Anti-PsbA Chicken Polyclonal Antibody
AGRIAS01016
: Agrisera
- Antibody type:Primary
- Antigen symbol:PSBA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:A. thaliana,H. vulgare
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- UniProtKB:A4QJR4
- Storage temperature:+4 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from available plant, algal and cyanobacterial PsbA sequences, including Arabidopsis thaliana
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PSBA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Chicken
Isotype:
Reactivity: A. thaliana, H. vulgare