Anti-TPTE2 Mouse Monoclonal Antibody [clone: 978CT6.1.4]
Catalog # ORIGTA324895
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TPTE2
- Clonality:Monoclonal
- Clone:978CT6.1.4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Format:Ascites
- Form:Liquid
- Storage buffer:crude ascites with 0,09% (W/V) sodium azide.
- Molecular weight:61.11 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Purified His-tagged TPIPb protein was used to produced this monoclonal antibody.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:5000
Type: Primary
Antigen: TPTE2
Clonality: Monoclonal
Clone: 978CT6.1.4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse