Anti-CBFA2T2 Rabbit Polyclonal Antibody
Catalog # ORIGTA324594
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CBFA2T2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:near N-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1 to 29
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:67.13 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This CBFA2T2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-29 amino acids from the N-terminal region of human CBFA2T2.
- Purification:purified by Protein chromatography, followed by peptide affinity purification
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000
Type: Primary
Antigen: CBFA2T2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human