Anti-CYP2D6 Rabbit Polyclonal Antibody
ORIGTA324552
: OriGene
- Antibody type:Primary
- Antigen symbol:CYP2D6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:near N-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:76 to 105
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:55.77 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This CYP2D6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 76-105 amino acids from the N-terminal region of human CYP2D6.
- Purification:purified by Protein chromatography, followed by peptide affinity purification
- Size:400 μl
- Pk:400 µl
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50
Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat