Anti-ARF6 Rabbit Polyclonal Antibody
Catalog # ORIGTA321390
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor 6
- Antigen symbol:ARF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:162 to 175
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:20 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to a region derived from 162-175 amino acids of Human ADP-ribosylation factor 6
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:2000-10000; Western Blot: 1:500-2000; Immunochemistry: 1:10-50
Type: Primary
Antigen: ARF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat