Anti-HIST1H3D Rabbit Polyclonal Antibody
ORIGTA322846
: OriGene
- Antibody type:Primary
- Antigen symbol:HIST1H3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:8 to 12
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:15 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Peptide sequence around aa.8~12 (R-K-S-T-G) derived from Human Histone H3.1.
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100-200
Type: Primary
Antigen: HIST1H3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat