Anti-IgG Rabbit Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
BSBTBA1092-1
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Rabbit
- Cross adsorption:No
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Rabbit
Isotype:
Reactivity: