Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Catalog # BSBTBA1058-0.5
Supplier:  Boster Bio
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Catalog # BSBTBA1058-0.5
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Cross adsorption:
    No
  • Size:
    500 μg
  • Pk:
    0,5 mg

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: