Order Entry
Slovenia
ContactUsLinkComponent
Anti- IgG Antibody (Unconjugated)
Anti- IgG Antibody (Unconjugated)
  BSBTBA1041-5
Anti- IgG Antibody (Unconjugated)
  BSBTBA1041-5

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    Unconjugated
  • Cross adsorption:
    No
  • Size:
    5 mg
  • Pk:
    5 mg

 

 

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: