Specifications
- Antibody type:Primary
- Antigen name:intercellular adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:BB2|CD54|P3.58
- UniProtKB:P05362
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:NSO-derived human ICAM1 recombinant protein(Position: Q28-E480).
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Intercellular adhesion molecule 1(ICAM1) detection. Tested with WB, IHC-P in Human.
CD54, also known as ICAM-1. Intercellular adhesion molecule-1(ICAM1) is a ligand for lymphocyte function-associated(LFA) antigens. Intercellular adhesion molecule-1(ICAM-1) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for LFA-1, a beta 2 leukocyte integrin. ICAM1 protein is the major human rhinovirus receptor. ICAM1 gene is mapped to human chromosome 19. In humans, lymphocyte adhesion to cells is mediated by the protein heterodimer CD11a/CD18(Leu-CAMa, LFA-1) and its ligand CD54(ICAM-1).
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Type: Primary
Antigen: SLC10A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat