Anti-Mucin 5AC Rabbit Antibody
Catalog # BSBTPA2303
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Mucin 5AC
- Antigen symbol:MUC5AC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:tracheobronchial|mucin-5 subtype AC|mucin-5AC|mucin 5|major airway glycoprotein|tracheobronchial mucin|gastric mucin|subtypes A and C|TBM|MUC-5AC|lewis B blood group antigen|tracheobronchial/gastric|leB
- Amino acid number:552 - 567
- Sequence:TMQLFMQLAPKLRGQT
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Mucin 5AC (552-567aa TMQLFMQLAPKLRGQT).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Mucin-5AC(MUC5AC) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: MUC5AC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human