Clinical chemistry analyser, Microlab 300
634-4021
- Communication interface:Centronics type parallel port, RS232 type serial port
- Description:Clinical chemistry analyser, Microlab 300
- Electrical:100 – 240 VAC nominal
- W×D×H:400×365×170 mm
- Weight:8,5 kg
- Pk:1
This semi-automated clinical chemistry analyser shows excellent performance for low volumes and low concentrations.
- Full size qwerty keyboard and display
- Reagent volume 30% less than most competing systems
- Superior optical design offers required accuracy under every lab condition
The unique sipper and pump system provides the measuring unit with the reaction mixture. Measurement takes place in a specially designed micro flowcell, which is thermostatted by Peltier elements. The measuring volume of only 30 μl contributes to savings in reagent use.