Order Entry
Slovenia
ContactUsLinkComponent
Brightfield cell counters, Cellometer®
Brightfield cell counters, Cellometer®
Catalog # NEXCCMAUTOT4PLUS
undefined
Brightfield cell counters, Cellometer®
Catalog # NEXCCMAUTOT4PLUS
Specifications
  • Model:
    Auto T4 Plus
  • Description:
    Brightfield cell counter
  • Cell size:
    > 5 µm
  • Pk:
    1
Specifications
About this item
These automated cell counters are designed for counting small cells.
  • Highly reproducible results can be saved and exported in Excel
  • Improved data quality, versatility and flexibility
  • Templates for hundreds of cell types and easy definition of types of cells
  • Brightfield technique, powerful optics, pointer and analysis software
Applications usually include insect, algae, yeast, cancer, blood cells and spores.
Delivery: Supplied with a box of counting chambers.