Anti-ZONA OCCLUDENS 1 alpha-plus Rabbit Polyclonal Antibody
Catalog # USBIZ0750-11
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Zona Occludens 1 alpha-plus
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:886 to 995
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:50; Flow Cytometry: 1:50; Western Blot: 1:50; Immunochemistry: 1:50
Type: Primary
Antigen: ZONA OCCLUDENS 1 alpha-plus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat