Anti-TLR4/MD-2 Complex Rat Monoclonal Antibody (Biotin) [clone: 6A641]
Catalog # USBIT8050-35A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TLR4/MD-2 Complex
- Antigen symbol:TLR4
- Clonality:Monoclonal
- Clone:6A641
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, containing 0,02% sodium azide and 0,1%BSA
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunohistology: 1:50; Flow Cytometry: 1:50
Type: Primary
Antigen: TLR4/MD-2 Complex
Clonality: monoclonal
Clone: 6A641
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse