Order Entry
Slovenia
ContactUsLinkComponent
Anti-SSEA-3 Rat Monoclonal Antibody [clone: 13A270]
Anti-SSEA-3 Rat Monoclonal Antibody [clone: 13A270]
  USBIS6610-04
Anti-SSEA-3 Rat Monoclonal Antibody [clone: 13A270]
  USBIS6610-04

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SSEA-3
  • Clonality:
    Monoclonal
  • Clone:
    13A270
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    4 to 8c
  • Storage buffer:
    PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    25 µg
  • Pk:
    25 µG

 

 

Type: Primary
Antigen: SSEA-3
Clonality: Monoclonal
Clone: 13A270
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse