Anti-SLC1A5 Rabbit Polyclonal Antibody
Catalog # USBIS5329-20C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 1 (neutral amino acid transporter), member 5
- Antigen symbol:Solute carrier family 1, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 15-20 ?g/ml
Type: Primary
Antigen: SLC1A5 (solute carrier family 1 (neutral amino acid transporter), member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human