Anti-PHD1-3, pan Rabbit Polyclonal Antibody
USBIP3373A-100
- Antibody type:Primary
- Antigen name:PHD1-3, pan
- Antigen symbol:PHD1/2/3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% BSA.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:10000-1:100000; Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: PHD1-3, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human