Anti-Parainfluenza Virus Type 3, Hemagluttinin Mouse Monoclonal Antibody [clone: 5E403]
USBIP3107-29
- Antibody type:Primary
- Antigen name:Parainfluenza Virus Type 3, Hemagluttinin
- Clonality:Monoclonal
- Clone:5E403
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by protein A chromatography or sequential differential precipitation
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200
Type: Primary
Antigen: Parainfluenza Virus Type 3, Hemagluttinin
Clonality: monoclonal
Clone: 5E403
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: