Anti-Oxytocin Rabbit Polyclonal Antibody
Catalog # USBIO8250-09
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Oxytocin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig
- Format:Serum
- Form:lyophilized powder
- Storage buffer:50-100ul sterile ddH2O. Dilute further
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:400-1:800
Type: Primary
Antigen: Oxytocin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig