Anti-Ly-6A/E Rat Monoclonal Antibody [clone: D7]
Catalog # USBIL7715-05X
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:LY6A/E
- Clonality:Monoclonal
- Clone:D7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 8.0 Low Endotoxin.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Ly-6A/E
Clonality: monoclonal
Clone: D7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse