Anti-Ly-6A/E Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: D7]
USBIL7715-05L
- Antibody type:Primary
- Antigen symbol:LY6A/E
- Clonality:Monoclonal
- Clone:D7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Flow Cytometry: 1:5-1:10;
Type: Primary
Antigen: Ly-6A/E
Clonality: monoclonal
Clone: D7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse