Anti-Leukocyte Immunoglobulin-like Receptor 2 Mouse Monoclonal Antibody [clone: 11C374]
USBIL2028-09
- Antibody type:Primary
- Antigen name:Leukocyte Immunoglobulin-like Receptor 2
- Antigen symbol:LILR2
- Clonality:Monoclonal
- Clone:11C374
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: Leukocyte Immunoglobulin-like Receptor 2
Clonality: monoclonal
Clone: 11C374
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human