Anti-ETR 3 Rabbit polyclonal antibody
Catalog # USBIE8825-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:ETR 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage buffer:PBS0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ETR 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse