Anti-CD91, 85kD Mouse Monoclonal Antibody [clone: 5F126]
Catalog # USBIC2441-96D-1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD91, 85kD
- Clonality:Monoclonal
- Clone:5F126
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Rabbit,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,05M sodium phosphate, 0,1M sodium chloride, 1mM EDTA, pH 6.6.
- Molecular weight:85 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Recommended dilution: Western Blot: 1-5 ?g/ml; Immunochemistry: 1-10 ?g/ml
Type: Primary
Antigen: CD91, 85kD
Clonality: Monoclonal
Clone: 5F126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rabbit