Anti-Cadherin, pan Rabbit monoclonal antibody unconjugated [clone: 7H11]
Catalog # USBIC0107-10Y
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cadherin, pan
- Clonality:Monoclonal
- Clone:7H11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,100ug/ml BSA, 50% glycerol and less than 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Cadherin, pan
Clonality: Monoclonal
Clone: 7H11
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat