Anti-BTLA Rabbit Polyclonal Antibody
Catalog # USBIB5001-61A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:B and T lymphocyte associated
- Antigen symbol:BTLA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,2% gelatin, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 3-5 ?g/ml
Type: Primary
Antigen: BTLA (B and T lymphocyte associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse