Anti-Bmi-1, IN Mouse Monoclonal Antibody [clone: 6A10]
Catalog # USBIB2185-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:BMI-1, IN
- Clonality:Monoclonal
- Clone:6A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.1-0.5 ?g/ml; Western Blot: 1-3?g/ml
Type: Primary
Antigen: Bmi-1, IN
Clonality: monoclonal
Clone: 6A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human