Anti-Calbindin-D Mouse Monoclonal Antibody [clone: CB-D7]
Catalog # USBI144256
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Calbindin-D
- Clonality:Monoclonal
- Clone:CB-D7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Calbindin-D
Clonality: monoclonal
Clone: CB-D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat