Anti-AHSA1 Mouse Monoclonal Antibody [clone: 4H9-D7]
Catalog # USBI136750
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Activator Of Heat Shock 90kDa Protein ATPase Homolog 1
- Antigen symbol:AHSA1
- Clonality:Monoclonal
- Clone:4H9-D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:AHA1
- Storage buffer:PBS, pH 7,2, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: AHSA1 (AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast))
Clonality: monoclonal
Clone: 4H9-D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse