Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-SLC7A2 Rabbit Polyclonal Antibody
  USBI136429
Anti-SLC7A2 Rabbit Polyclonal Antibody
  USBI136429

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
  • Antigen symbol:
    Solute carrier family 7, member 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

  • Recommended dilution: Immunochemistry: 15 μg/ml

Type: Primary
Antigen: SLC7A2 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human