Order Entry
Slovenia
ContactUsLinkComponent
Anti-CWC22 Mouse Polyclonal Antibody
Anti-CWC22 Mouse Polyclonal Antibody
Catalog # USBI128802
Supplier:  US Biological
Anti-CWC22 Mouse Polyclonal Antibody
Catalog # USBI128802
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CWC22 spliceosome-associated Protein homolog (S. cerevisiae)
  • Antigen symbol:
    CWC22
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CWC22 (CWC22 spliceosome-associated protein homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human