Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-DOCK4 Mouse Monoclonal Antibody [clone: 3E7]
  USBI125985
Anti-DOCK4 Mouse Monoclonal Antibody [clone: 3E7]
  USBI125985

 

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 4
  • Antigen symbol:
    DOCK4
  • Clonality:
    Monoclonal
  • Clone:
    3E7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1867 to 1966
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    NQVNEQSAPLPVPVPVPVPSYGGEEPVRKESKTPPPYSVYERTLRRPVPLPHSLSIPVTSEPPALPPKPLAARSSHLENGARRTDPGPRPRPLPRKVSQL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: DOCK4 (dedicator of cytokinesis 4)
Clonality: monoclonal
Clone: 30000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human