Anti-CTBS Mouse Monoclonal Antibody [clone: 1B5-1B9]
Catalog # USBI125434
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chitobiase, di-N-acetyl-
- Antigen symbol:CTBS
- Clonality:Monoclonal
- Clone:1B5-1B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:DCPCPEPELCRPIRHHPDFEVFVFDVGQKTWKSYDWSQITTVATFGKYDSELMCYAHSKGARVVLKGNL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CTBS (chitobiase, di-N-acetyl-)
Clonality: monoclonal
Clone: 1B5-1B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human