Order Entry
Slovenia
ContactUsLinkComponent
Anti-SOD1 (g93A mutant) Mouse Monoclonal Antibody [clone: B8H10]
Anti-SOD1 (g93A mutant) Mouse Monoclonal Antibody [clone: B8H10]
Catalog # MEDMMM-0070-P
Supplier:  MédiMabs
Anti-SOD1 (g93A mutant) Mouse Monoclonal Antibody [clone: B8H10]
Catalog # MEDMMM-0070-P
Supplier:  MédiMabs

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SOD1 g93A mutant
  • Antigen symbol:
    SOD1
  • Clonality:
    Monoclonal
  • Clone:
    B8H10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: SOD1 (g93A mutant)
Clonality: monoclonal
Clone: B8H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human