Anti-CFL1 Rabbit Polyclonal Antibody
Catalog # USBI034073
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cofilin 1
- Antigen symbol:CFL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser3
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Dot blot 1:100-500
Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human