Anti-Actin, pan Mouse Monoclonal Antibody [clone: 13B812]
Catalog # USBI031348
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:actin, pan
- Clonality:Monoclonal
- Clone:13B812
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:500-1:1000; Western Blot: 1:5000-1:10000
Type: Primary
Antigen: Actin, pan
Clonality: Monoclonal
Clone: 13B812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat