Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Anti-SOX2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 57CT23.3.4]

Anti-SOX2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 57CT23.3.4]

100 µl
Catalog # USBI031255-FITC
Supplier:   US Biological
Anti-SOX2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 57CT23.3.4]
Catalog # USBI031255-FITC
Supplier:  US Biological
Anti-SOX2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 57CT23.3.4]
Anti-SOX2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 57CT23.3.4]

100 µl

Price
929,00 €
Log in for availability
1 * 200 µl
Product Details & Documents

Recommended dilution: FLISA: 1:10000; Western Blot: 1:200-2000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Monoclonal
Clone: 57CT23.3.4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Documents

Specifications
  • Size:
    100 µl
  • Western blot:
    Yes
  • Clonality:
    Monoclonal
  • Flow cytometry:
    Yes
  • Purification:
    purified by Protein G chromatography
  • Antibody type:
    Primary
  • Form:
    liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Storage temperature:
    Cool
  • Host:
    Mouse
  • Shipping temperature:
    Ice
  • Antigen name:
    SRY (sex determining region Y)-box 2
  • Cat. No.:
    USBI031255-FITC
  • Reactivity:
    Human
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Antigen symbol:
    SOX2
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Clone:
    57CT23.3.4
  • ELISA:
    Yes