Anti-AIF1 Mouse Monoclonal Antibody [clone: 13A155]
Catalog # USBI029413
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Allograft Inflammatory Factor 1
- Antigen symbol:AIF1
- Clonality:Monoclonal
- Clone:13A155
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:300
Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: Monoclonal
Clone: 13A155
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat