Anti-AKT1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 5E5.F5.D7]
Catalog # ROCK200-302-J34
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:v-akt murine thymoma viral oncogene homolog 1
- Antigen symbol:AKT1
- Clonality:Monoclonal
- Clone:5E5.F5.D7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:207
- Antigen synonyms:CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
- Storage buffer:0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-AKT1 Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 Protein followed by monoclonal development.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Anti-AKT1 FITC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 5E5.F5.D7
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human