Anti-CUL5 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14C5]
Catalog # ROCK100-401-A05
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:cullin 5
- Antigen symbol:CUL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:8065
- Antigen synonyms:VACM-1|VACM1
- Amino acid number:4 to 18
- Storage buffer:0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 4-18 of Human Cul5 (N-terminus) coupled to KLH.
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This antibody reacts with human Cul5 by western blot and immunoprecipitation. The antibody immunoprecipitates Protein from cell lysates (using 293T, U2OS and others). This antibody also co-immunoprecipitates associated Proteins.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CUL5
Clonality: Monoclonal
Clone: 14C5
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human