Anti-Loki / Chk2 Goat Polyclonal Antibody
EVBIEB11423
- Antibody type:Primary
- Antigen symbol:LOKI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Drosophila
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:Chk2
- Amino acid number:115 to 128
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Loki / Chk2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Drosophila