Anti-DEFB107A / DEFB107B Goat polyclonal antibody unconjugated
EVBIEB11110
- Antibody type:Primary
- Antigen symbol:DEFB107A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:DEFB107B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DEFB107A / DEFB107B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human