Anti-DAO Goat polyclonal antibody unconjugated
EVBIEB11100
- Antibody type:Primary
- Antigen name:D-amino-acid oxidase
- Antigen symbol:DAO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:286 to 298
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DAO (D-amino-acid oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: