Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-CRLS1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB11070
Supplier:  Everest Biotech
undefined
Anti-CRLS1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB11070
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cardiolipin synthase 1
  • Antigen symbol:
    CRLS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: CRLS1 (cardiolipin synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: